Endoproteinase Lys-C, sequencing grade, Lysobacter enzymogenes
Lieferant: Calbiochem (Part of Merck)
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
324715-5
324715-5EA 340 EUR InStock
324715-5
Endoproteinase Lys-C, sequencing grade, Lysobacter enzymogenes
Protein-Extraktion Protein Extraction Kits
Native endoproteinase Lys-C from Lysobacter enzymogenes. Serine protease that specifically hydrolyses amide and peptide ester bonds at the carboxylic side of lysine in peptides and proteins. Designed for protein sequencing or sequence verification, analysis of protein structural domains, and cleavage of fusion proteins. Inhibited by aprotinin, DFP, leupeptin, and TLCK. Suggested working concentration: 1:20 to 1:100 (protease: protein by weight) for sequence analysis.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge