Pesticide reference standard, Isotianil, PESTANAL®, analytical standard, Supelco®
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
32524-100MG
32524-100MGEA 101 EUR InStock
32524-100MG
Pesticide reference standard, Isotianil, PESTANAL®, analytical standard, Supelco®
Standards Organische Standards
Isotianil has been used as a reference standard for the determination of isotianil in brown rice and soil using supercritical fluid extraction and gas chromatography/mass spectrometry method.
  • Helps in preventing the disease like rice blast, caused by rice blast fungi
Commonly used pesticide which activates defence responses in plants against a wide range of pathogens.
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f2dd7ece3dba2d0d795b8668d0cea5efa0187c6&vid=5f2dd7ece3dba2d0d795b8668d0cea5efa0187c6&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge