Supelco® PESTANAL® Organic Reference Standard, Spirotetramat
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
32713-100MG
32713-100MGEA 161 EUR InStock
32713-100MG
Supelco® PESTANAL® Organic Reference Standard, Spirotetramat
Standards Organische Standards
  • GHS signal: GHS07
Spirotetramat may be used as a reference standard for QuEChERS based methodology for the determination of spirotetramat and its relevant metabolites in pistachio samples by liquid chromatography-tandem mass spectrometry.
Achtung! Bitte beachten: P201 - P273 - P280 - P302 + P352 - P305 + P351 + P338 - P308 + P313
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73bde3803cb7c07d33d3729a333eb6063ac40a4f&vid=73bde3803cb7c07d33d3729a333eb6063ac40a4f&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge