Organic reference standard, Aflatoxin B2, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
32755-5MG
32755-5MGEA 1730 EUR InStock
32755-5MG
Organic reference standard, Aflatoxin B2, Supelco®
Standards Organische Standards
Aflatoxins are mycotoxins that are generated as a result of secondary metabolism of various fungal species such as AspergillusPenicillium and Fusarium.
Aflatoxin B₂ can be used as a reference standard for the determination of aflatoxin B₂ in animal feed by high performance liquid chromatography with fluorescence detection (HPLC-FLD), as well as quick, easy, cheap, effective, rugged and safe (QuEchERS) extraction method combined with ultra-high performance liquid chromatography-high resolution mass spectrometry (UHPLC-HRMS), in eggs by QuEchERS extraction method followed by analysis using UHPLC with tandem mass spectrometry (MS/MS) operating on electrospray-ionization interface (ESI) mode, in Brazil nuts by immunoaffinity column (IAC) and clean-up HPLC-FLD and in plant-based milk and dairy products by dispersive liquid-liquid microextraction (DLLME), QuEchERS extraction method and HPLC-FLD.
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1585fbecca8117b2e77b97f7adc5bcc64d2fa8bd&vid=1585fbecca8117b2e77b97f7adc5bcc64d2fa8bd&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge