Organic reference standard, Aflatoxin B1-13C17 solution, analytical standard, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
32764-1ML
32764-1MLEA 2600 EUR InStock
32764-1ML
Organic reference standard, Aflatoxin B1-13C17 solution, analytical standard, Supelco®
Standards Organische Standards
Aflatoxin B₁-¹³C₁₇ is the ¹³C isotope-labeled analog of the mycotoxin, aflatoxin.
It may be used as an analytical standard for the determination of aflatoxin B₁ in food grade gums, Pu-erh tea samples, animal feed as well as milk, milk-based infant formula, and feed samples by stable isotope dilution and liquid chromatography combined with tandem mass spectrometry (LC-MS/MS).
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24b7f0cf3467de0b047e8dd8de9533a21f9759b6&vid=24b7f0cf3467de0b047e8dd8de9533a21f9759b6&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge