Organic reference standard, Leucocrystal Violet-d6, analytical standard, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
32834-10MG
32834-10MGEA 507 EUR InStock
32834-10MG
Organic reference standard, Leucocrystal Violet-d6, analytical standard, Supelco®
Standards Organische Standards
Leucocrystal violet-d₆ might be used as an internal standard in the determination of triphenylmethane dyes in rainbow trout samples using liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS).
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcc197898fcfc3e1cc302f78fc363ca825d4f6c5&vid=dcc197898fcfc3e1cc302f78fc363ca825d4f6c5&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge