Fish spine beads, AL23
Lieferant: Alfa Aesar
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
32905.KX 32906.KX 32907.KX 32908.KX
32905.KXEA 145 EUR InStock
32905.KX 32906.KX 32907.KX 32908.KX
Fish spine beads, AL23
Kugeln Keramikkugeln
Pure Al₂O₃ Aluminium oxide, AL 23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge