Crucibles, Tammann, aluminum oxide
Lieferant: Alfa Aesar
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
33052.KF 33053.KF 33055.KF 33056.KF 33058.KF
33052.KFEA 43.9 EUR InStock
33052.KF 33053.KF 33055.KF 33056.KF 33058.KF
Crucibles, Tammann, aluminum oxide
Tiegel
These crucibles according to Tammann are manufactured from aluminium oxide. They display superior characteristics both at elevated temperature and at high frequencies, and are resistant to corrosive liquids and gases. Oxide ceramics are also inert to oxidation and are not subject to radiation damage.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge