Bottles, wide neck, HDPE, economy, with screw cap, Nalgene®
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
DS2185-0004 DS2185-0016 DS2185-0032 DS2185-0008
331-4213EA 154 EUR InStock
331-4213 331-4215 NALGDS2185-0032 331-0300
Bottles, wide neck, HDPE, economy, with screw cap, Nalgene®
Flaschen Kunststoffflaschen
Wide mouth amber bottles made up of HDPE designed to protect the light sensitive liquids. It is used for storage, shipping and packaging.
  • Economical and recyclable
  • Designed for single use
  • Chemical resistance to most acids, bases and alcohols
  • Can be used in freezer to −100 °C for long term storage
  • Linerless caps ensure leakproof performance
These bottles are for single use and non autoclavable.
Zertifizierungen: Compliance with U.S. Pharmacopoeia 23 requirements for maximum light transmission.
Lieferumfang: Bottles are supplied with amber polypropylene screw closures.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge