33241.22
33241.36
33241.A4
33241.22EA
38.4
EUR
InStock
33241.22
33241.36
33241.A4
L(+)-Kaliumnatriumtartrat Tetrahydrat 99,0-102,0% ACS
L(+)-Kaliumnatriumtartrat Tetrahydrat
Purity: 99.0-102.0%
CAS No.: 6381-59-5
Molecular Formula: C4H4KNaO6.4H2O
Formula Weight: 282.23 (210.29anhy)
Physical Form: Crystalline
Density: 1.77
pH: 6.0-8.5
Melting Point: 70-80°
Beilstein Reference: 6113568
Merck Reference: 14,7670
MDL No.: MFCD00150989