33358.22
33358.36
33358.A3
33358.22EA
38
EUR
InStock
33358.22
33358.36
33358.A3
Kaliumhexacyanoferrat(II) Trihydrat 98,5-102,0% ACS
Kaliumhexacyanoferrat(II) Trihydrat
Purity: 98.5-102.0%
CAS No.: 14459-95-1
Molecular Formula: K4Fe(CN)6.3H2O
Formula Weight: 422.39 (368.36anhy)
Physical Form: Crystalline
Density: 1.85
Appearance: Pale yellow
Odour: Odourless
pH: 9.5
Melting Point: 69-71°
Merck Reference: 14,7631
MDL No.: MFCD00167023