Organic reference standard, Aflatoxin Mix 4 solution, analytical standard, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
33415-2ML
33415-2MLEA 894 EUR InStock
33415-2ML
Organic reference standard, Aflatoxin Mix 4 solution, analytical standard, Supelco®
Standards Organische Standards
Verpackung: B1, G1, B2, G2 in acetonitrile (20 ug/ml each).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge