33480.01
33480.03
33480.06
33480.01EA
119
EUR
InStock
33480.01
33480.03
33480.06
Osmium(III)chlorid Trihydrat ≥99,99% (Metall-Basis) Os 52-56%, Premion®
Osmium(III)chlorid Trihydrat
Purity: 99.99%
CAS No.: 135296-80-9
Molecular Formula: Cl3Os.3H2O
Formula Weight: 350.60 (296.56anhy)
Physical Form: Crystalline
Appearance: Red-black
Melting Point: dec.
UN No.: 3260
MDL No.: MFCD00011148