pH buffer solutions, referenced at 20 °C, traceable to SRM from NIST, Fluka™
Lieferant: HONEYWELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
33544-1L 33545-1L 33547-1L 33643-10L-VP 33646-10L-VP 33648-10L-VP 33646-500ML 33643-100ML 33643-5L-VP 33646-100ML 33646-5L-VP 33648-500ML 33643-1L 33643-500ML 33643-6X1L 33646-1L 33646-6X1L 33648-1L 33648-6X1L 31044-1L 31044-6X1L 31045-1L 31045-6X1L 31046-1L 31046-6X1L 33544-6X1L 33545-6X1L 33547-6X1L 31045-100ML
33544-1LEA 57.96 EUR InStock
33544-1L 33545-1L 33547-1L 33643-10L-VP 33646-10L-VP 33648-10L-VP 33646-500ML 33643-100ML 33643-5L-VP 33646-100ML 33646-5L-VP 33648-500ML 33643-1L 33643-500ML 33643-6X1L 33646-1L 33646-6X1L 33648-1L 33648-6X1L 31044-1L 31044-6X1L 31045-1L 31045-6X1L 31046-1L 31046-6X1L 33544-6X1L 33545-6X1L 33547-6X1L 31045-100ML
pH buffer solutions, referenced at 20 °C, traceable to SRM from NIST, Fluka™
Puffer pH-Puffer
  • Citrate and phosphate buffer solutions contain fungicide
  • Traceable to SRM from NIST and PTB
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge