Organic reference standard, Deoxynivalenol-d1 solution, analytical standard, Supelco®
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
34155-2ML
34155-2MLEA 515 EUR InStock
34155-2ML
Organic reference standard, Deoxynivalenol-d1 solution, analytical standard, Supelco®
Standards Organische Standards
Deoxynivalenol-d1 solution can be used as working standard in the analysis of trichothecenes and zearalenones in grain cereal, flour and bread using liquid chromatography coupled with mass spectrometry (LC/MS-MS). It can also be used as an internal standard in the determination of mycotoxins in baby foods using liquid chromatography coupled with mass spectrometry (LC/MS-MS).
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=756701c0310e3ac9bf69bbc06ebf7f6b4c029c50&vid=756701c0310e3ac9bf69bbc06ebf7f6b4c029c50&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge