Pesticide reference standard, Ronidazole-d3, VETRANAL(TM), analytical standard, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
34217-10MG
34217-10MGEA 475 EUR InStock
34217-10MG
Pesticide reference standard, Ronidazole-d3, VETRANAL(TM), analytical standard, Supelco®
Standards Organische Standards
Ronidazole-d₃ may be used internal standard in the determination of avermectins, benzimidazoles and nitroimidazoles residues in bovine muscle tissue using ultra-high performance liquid chromatography coupled with mass spectrometry (UHPLC/MS-MS).
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46f80e34b69569000fb6c19386f11cb781faa84e&vid=46f80e34b69569000fb6c19386f11cb781faa84e&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge