Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

D(-)-Sorbit 91,0-100,5% (durch Trockensubstanz) NF, Multikompendial, J.T.Baker®

Lieferant: AVANTOR PERFORMANCE MATERIALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


Veränderungsmanagement-Kategorie= R

3449-09EA 4630 EUR InStock
3449-09 3449-39
D(-)-Sorbit 91,0-100,5% (durch Trockensubstanz) NF, Multikompendial, J.T.Baker®
D(-)-Sorbit
Only class 2 solvents (ethylene glycol) are likely to be present. All are below option 1 limits.


Formel: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
Siedepunkt: 295
MDL Number: MFCD00004708
CAS-Nummer: 50-70-4
Merck Index: 13,08797
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82054e8cfed2cf3e8b739c3cb816c4f91bd2735&vid=f82054e8cfed2cf3e8b739c3cb816c4f91bd2735&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets N.F. Requirements
GMP Manufactured Product
Meets J.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical 91.0 - 100.5 %
NF - Assay (anhydrous basis) 91.0 - 100.5 %
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - pH of 10% solution 3.5 - 7.0
NF - Reducing Sugars ≤ 0.3 %
NF - Residue on Ignition ≤ 0.1 %
NF - Nickel (Ni) ≤ 1 ppm
NF - Chloride (Cl) ≤ 0.0050 %
NF - Sulfate (SO₄) ≤ 0.01 %
NF - Water ≤ 1.5 %
NF - Total Aerobic Microbial Count, cfu/g ≤ 1000
NF - Total Yeast and Mold Count, cfu/g ≤ 100
EP/BP - Assay (anhydrous basis) 97.0 - 102.0 %
EP/BP - Identification A Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Conductivity, uS cm-1 ≤ 20
EP/BP - Reducing Sugars ≤ 0.2 %
EP/BP - Related Substances - Any impurity, each ≤ 2.0 %
EP/BP - Related Substances - Total ≤ 3.0 %
EP/BP - Water ≤ 1.5 %
Total Viable Aerobic Count - Total Aerobic Microbial Count, cfu/g ≤ 100
Total Viable Aerobic Count - Total Yeast and Mold Count, cfu/g ≤ 100
Total Viable Aerobic Count - Escherichia coli (negative) Passes Test
Salmonella (negative) Passes Test
Endotoxin Concentration, <2.5 IU/g Passes Test
JP - Assay (dried basis) ≥ 97.0 %
JP - Identification A Passes Test
JP - Identification B Passes Test
JP - Identification C Passes Test
JP - Clarity and Color Passes Test
JP - Acidity or Alkalinity Passes Test
JP - Chloride (Cl) ≤ 0.005 %
JP - Sulfate (SO₄) ≤ 0.006 %
JP - Heavy Metals (as Pb) ≤ 5 ppm
JP - Nickel (Ni) Passes Test
JP - Arsenic (As) ≤ 1.3 ppm
JP - Glucose Passes Test
JP - Sugars Passes Test
JP - Loss on Drying ≤ 2.0 %
JP - Residue on Ignition ≤ 0.02 %
Only Class 2 solvents (Ethylene Glycol) are likely to be present. All are below Option 1 limits.

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge