34493-2.5L
34493-2.5LEA
210.18
EUR
InStock
34493-2.5L
Hexan (Isomerengemisch), CHROMASOLV™ für die Analyse von Pestizidrückständen, Riedel-de Haën™
Hexan (Isomerengemisch)
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent