Natriumhydroxid 2 M NaOH (2N) volumetrische Lösung, Fluka™
Lieferant: HONEYWELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
Gefahr
35254-1L
35254-1LEA 67.07 EUR InStock
35254-1L
Natriumhydroxid 2 M NaOH (2N) volumetrische Lösung, Fluka™
Sodium hydroxide
Honeywell's high-quality titration products cover the complete titration workflow and include:
  • Solutions for volumetric titration (acids, bases, or salts)Buffers available as concentrates or ready-to-useReagents for complexometry:Aminopolycarboxylic acids (EDTA/NTA analogs)
  • Masking agents
  • Indicators for volumetric and complexometric titrations
UNSPSC Code: 12352316
Formel: NaOH
Siedepunkt: 105…140 °C (1,013 hPa)
Melting Pt: –12 – –10 °C,
Dichte: 1,08 g/cm³ (20 °C)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00003548
CAS-Nummer: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II
Order Now
Spezifikation Testergebnisse
Factor 0.999 - 1.001
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge