35635.K2
35635.K2EA
48.8
EUR
InStock
35635.K2
Natriumhydroxid standardisierte Lösung 10.0 N in wässriger Lösung
Sodium hydroxide
CAS No.: 1310-73-2
Molecular Formula: NaOH
Formula Weight: 40.00
Physical Form: Liquid
Density: 1.327
Appearance: Colourless
Odour: Odourless
UN No.: 1824
MDL No.: MFCD00003548