Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Human processed serum (HPS), TSH depleted

Lieferant: SEQENS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

36000D-100ML 36000D-500ML 36000D-1L
36000D-100MLEA 397 EUR InStock
36000D-100ML 36000D-500ML 36000D-1L
Human processed serum (HPS), TSH depleted
Seren
SEQENS IVD manufactures TSH Depleted Human Processed Serum.

  • Dry extract: 65 to 85 g/L
  • Total proteins: 55 to 75 g/L
  • TSH: <0,1 UI/L
  • Filtered through a 0,2 µm membrane
  • Shelf life: 2 years

For this product, source material has been tested and found negative or non-reactive (FDA approved and/or CE marked test methods) for anti-HIV 1+2 and anti-HCV antibodies, HBs antigen, HIV 1 and HCV RNA and Syphilitic serology.

Complete donor traceability on file.

Achtung! Bitte beachten: Only for research or manufacture of in vitro diagnostic reagents. Not for therapeutic use. This product of human origin should be considered as potentially infectious and handled accordingly.

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge