36290.06
36290.14
36290.22
36290.06EA
34.6
EUR
InStock
36290.06
36290.14
36290.22
4-Amino-3-hydroxy-1-naphthalinsulfonsäure ≥90% ACS
4-Amino-3-hydroxy-1-naphthalenesulphonic acid
Purity: 90%
CAS No.: 116-63-2
Molecular Formula: C10H9NO4S
Formula Weight: 239.25
Physical Form: Powder
Melting Point: ca 295° dec.
Beilstein Reference: 2697469
Merck Reference: 14,454
MDL No.: MFCD00004019