36393.36
36393.36EA
75
EUR
InStock
36393.36
Blei(II)acetat basisch ACS
Blei(II)acetat basisch
CAS No.: 1335-32-6
Molecular Formula: C4H6O4Pb.2Pb(OH)2
Formula Weight: 807.71
Physical Form: Powder
Sensitivity: Air Sensitive
Merck Reference: 14,5419
MDL No.: MFCD00013041