36626.14
36626.22
36626.36
36626.14EA
51.3
EUR
InStock
36626.14
36626.22
36626.36
3,3'-Dimethylbenzidin Dihydrochorid ACS
3,3'-Dimethylbenzidin Dihydrochorid
CAS No.: 612-82-8
Molecular Formula: C14H16N2.2HCl
Formula Weight: 285.22
Physical Form: Solid
Sensitivity: Air and Light Sensitive
Appearance: White to cream
Odour: Odourless
Melting Point: 300°
UN No.: 3077
MDL No.: MFCD00012960