Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Natriumhydroxid 50% in wässriger Lösung, CMOS für Mikroelektronik, J.T.Baker®

Lieferant: AVANTOR PERFORMANCE MATERIALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

3719-09EA 0 EUR InStock
3719-09
Natriumhydroxid 50% in wässriger Lösung, CMOS für Mikroelektronik, J.T.Baker®
Sodium hydroxide


Formel: NaOH
MW: 40 g/mol
Siedepunkt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Dichte: 1.53 g/cm³ (20 °C)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00003548
CAS-Nummer: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627
Order Now
Spezifikation Testergebnisse
For Microelectronic Use 50.0 - 52.0 %
Protect from air to avoid absorption of carbon dioxide. = 10
Store at temperatures above 60°F (15°C) = 30 ppm
Assay (NaOH) (by acidimetry) 49.5 - 51.5 %
Color (APHA) ≤10
Insoluble Matter ≤30 ppm
Sodium Carbonate (Na₂CO₃) ≤0.05 %
Ammonium Hydroxide Precipitate ≤0.005 %
Chloride (Cl) ≤20 ppm
Heavy Metals (as Ag) ≤2 ppm
Nitrogen Compounds (as N) ≤1 ppm
Phosphate (PO₄) ≤1 ppm
Trace Impurities - Potassium (K) ≤50 ppm
ACS - Sulfate (SO₄) ≤5 ppm
Trace Impurities - Aluminum (Al) ≤1000.0 ppb
Trace Impurities - Calcium (Ca) ≤1500.0 ppb
Trace Impurities - Chromium (Cr) ≤500.0 ppb
Trace Impurities - Copper (Cu) ≤100.0 ppb
Trace Impurities - Iron (Fe) ≤1000.0 ppb
Trace Impurities - Lead (Pb) ≤300.0 ppb
Trace Impurities - Magnesium (Mg) ≤500.0 ppb
Trace Impurities - Mercury (Hg) ≤50 ppb
Trace Impurities - Nickel (Ni) ≤100.0 ppb
Trace Impurities - Silicon (Si) ≤2000.0 ppb
Trace Impurities - Zinc (Zn) ≤300.0 ppb
Store above 58°F (12°C)
For additional information, go to www.askavantor.com. Search keywords
freezing" and product name."

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge