4-Methoxybiphenyl 97%, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
374776-5G
374776-5GEA 79.7 EUR InStock
374776-5G
4-Methoxybiphenyl 97%, Sigma-Aldrich®
4-Methoxybiphenyl
4-Methoxybiphenyl has been used as a standard reagent whose fluorescence intensity is associated with the fluorescence characteristics of the products of derivatisation reaction for aryl halides with phenylboronic acid (PBA).
Formel: C₁₃H₁₂O
MW: 184.24 g/mol
Melting Pt: 86...90 °C (Lit.)
Storage Temperature: Room Temperature
MDL Number: MFCD00014897
CAS-Nummer: 613-37-6
EINECS: 210-339-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c986c6b605d043526b8d528f1ea930bcf8df611&vid=9c986c6b605d043526b8d528f1ea930bcf8df611&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Appearance White to off-white powder, crystals, crystalline powder and/or chunks
Infrared spectrum Conforms to structure
Gas liquid 96.5% (minimum)
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge