Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

di-Natriumhydrogenphosphat Heptahydrat 98,0-100,5% (Trockenmasse) USP, J.T.Baker®

Lieferant: AVANTOR PERFORMANCE MATERIALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

Veränderungsmanagement-Kategorie= HR

3803-01EA 143 EUR InStock
3803-01 3803-07 3803-R 3803-65
di-Natriumhydrogenphosphat Heptahydrat 98,0-100,5% (Trockenmasse) USP, J.T.Baker®
di-Natriumhydrogenphosphat Heptahydrat


Formel: HNa₂O₄P·7H₂O
MW: 268.07 g/mol
Melting Pt: 48 °C
Dichte: 1.7 g/cm³ (20 °C)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00149180
CAS-Nummer: 7782-85-6
EINECS: 231-448-7
Order Now
Spezifikation Testergebnisse
GMP Manufactured Product 98.0 - 100.5%
Meets U.S.P Requirements ≤16 ppm
CAUTION: For Manufacturing, processing or repackaging ≤0.06%
Bulk Pharmaceutical Chemical ≤2.5
Appearance (Colorless or translucent crystals or granules) Passes Test
USP - Assay (Na₂HPO₄) (dried basis) 98.0 - 100.5 %
USP - Arsenic (As) ≤ 16 ppm
USP - Chloride (Cl) ≤ 0.06 %
USP - Endotoxin Concentration (EU/g) ≤ 2.5
USP - Identification Passes Test
USP - Insoluble Substances ≤ 0.4 %
USP - Loss on Drying 43.0 - 50.0 %
USP - Sulfate (SO₄) ≤ 0.2 %
EP - Identification A Passes Test
EP - Identification C Passes Test
EP - Identification D Passes Test
EP - Reducing Substances Passes Test
EP - Monosodium Phosphate ≤ 2.5 %
EP - Chloride (Cl) ≤ 200 ppm
EP - Sulfate (SO₄) ≤ 500 ppm
EP - Appearance of Solution Passes Test
EP - Iron (Fe) ≤ 20 ppm
EP - Assay (dried basis) 98.0 - 101.0 %
JP - Arsenic (As) ≤ 2 ppm
JP - Assay (Na₂HPO₄) (dried basis) 98.0 - 101.0 %
JP - Carbonate (CO₃) Passes Test
JP - Chloride (Cl) ≤ 0.014 %
JP - Clarity and Color of Solution Passes Test
JP - Heavy Metals ≤ 10 ppm
JP - Identification Passes Test
JP - pH (1 in 50) 9.0 - 9.4
JP - Sulfate (SO₄) ≤ 0.038 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
The EP results are derived from testing Sodium Phosphate, Dibasic,
7-Hydrate, U.S.P. to the EP chemical specifications derived from
Disodium Phosphate Dihydrate and Disodium Phosphate Dodecahydrate.
The JP results are derived from testing Sodium Phosphate, Dibasic,
7-Hydrate, U.S.P. to the JP chemical specifications derived from Dibasic
Sodium Phosphate Hydrate.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge