Petri dish holder
Lieferant: SciLabware
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SWP062
391-1150EA 30 EUR InStock
391-1150
Petri dish holder
Gestelle Spender für Petrischalen
Polyamide epoxy coated steel. Holds up to six 90 mm petri dishes.
  • Better heat and chemical resistance than other coatings
  • Ideal for small batch incubation
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge