Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Bacterial cell spreaders, SP Bel-Art

Lieferant: Bel-Art Products, a Part of SP

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

F377360009 F377360006
391-2007EA 47.8 EUR InStock
391-2007 391-2006
Bacterial cell spreaders, SP Bel-Art
Schaber Zellschaber
For spreading cells onto the surface of an agar plate to form individual colonies. These stainless spreaders have a smooth triangular end with a straight spreading bar.

  • The small size is ideal for 6 cm plates and the large size for 9 cm plates
  • Autoclavable and may be flame sterilised

Overall length: 203 mm

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge