Contact plate, Sterilin®
Lieferant: Sterilin
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
504
391-2031EA 716 EUR InStock
391-2031
Contact plate, Sterilin®
Zellkulturschalen Petrischalen
Ideal for use in routine hygiene monitoring of surfaces.
  • Concave profile of the dish raises the profile of the media when set giving better contact between the agar bed and surface under test
  • Aseptically manufactured under cleanroom conditions (class 7 ISO 14644) to exclude microbiological contamination
  • Triple vented to aid gaseous exchange
  • Numbered grid on the base facilitates colony counting
  • Deep skirted base aids stability when stacked
Dimensions shown are for outer Ø base and outer height (mm).
Zertifizierungen: Conforms to IES 1993 and IPF 1990 standards.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge