Zellkulturröhrchen, CELLSTAR®
Lieferant: GREINER BIO ONE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
163160_1000
391-3410EA 938 EUR InStock
391-3410
Zellkulturröhrchen, CELLSTAR®
Röhrchen Kulturröhrchen
PS, pre-treated, sterile.
  • Physical surface treatment for improved cell adhesion
  • Available with screw cap, bayonet cap or two position vented stopper
All tubes are certified free of RNase, DNase and human DNA, are non-pyrogenic and non-cytotoxic.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge