39198.AK
39198.K2
39198.M1
39198.AKEA
43.5
EUR
InStock
39198.AK
39198.K2
39198.M1
1-Chlorbutan ≥99,5% für die HPLC
1-Chlorbutan
Purity: 99.5+%
CAS No.: 109-69-3
Molecular Formula: CH3(CH2)3Cl
Formula Weight: 92.57
Flash Point: -12°(10°F)
Physical Form: Liquid
Density: 0.886
Refractive Index: 1.4024
Odour: Pungent
Melting Point: -123°
Boiling Point: 77-78°
Beilstein Reference: 1730909
Merck Reference: 14,1560
UN No.: 1127
MDL No.: MFCD00001009