39305.03
39305.06
39305.14
39305.03EA
43.6
EUR
InStock
39305.03
39305.06
39305.14
Quecksilber(II)trifluormethansulfonat ≥98%
Quecksilber(II)trifluormethansulfonat
Purity: 98%
CAS No.: 49540-00-3
Molecular Formula: C2F6HgO6S2
Formula Weight: 498.72
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: White
Melting Point: 350°
UN No.: 2025
MDL No.: MFCD00144746