39434.09
39434.18
39434.09EA
103
EUR
InStock
39434.09
39434.18
Potassium tetracyanonickelate(II) hydrate
Potassium tetracyanonickelate(II) hydrate
CAS No.: 339527-86-5
Molecular Formula: K2[Ni(CN)4].xH2O
Formula Weight: 240.99(anhy)
Physical Form: Crystalline
Density: 1.875
Melting Point: 100° -H2O
Solubility: Soluble in water
UN No.: UN1588
MDL No.: MFCD00049669