39453.06
39453.14
39453.06EA
57.7
EUR
InStock
39453.06
39453.14
Nickel(II) phthalocyanine
Nickel(II) phthalocyanine
CAS No.: 14055-02-8
Molecular Formula: Ni(C32H16N8)
Formula Weight: 571.24
Physical Form: Powder
Appearance: Dark blue to purple
Melting Point: 300°
MDL No.: MFCD00041954