39502.14
39502.22
39502.14EA
242
EUR
InStock
39502.14
39502.22
Kaliumorthovanadat ≥99,9% (Metall-Basis)
Potassium orthovanadate
Purity: 99.9%
CAS No.: 14293-78-8
Molecular Formula: K3VO4
Formula Weight: 232.24
Physical Form: Granular
Odour: Odourless
UN No.: 3285
MDL No.: MFCD00210642