39554.36
39554.A3
39554.36EA
98.8
EUR
InStock
39554.36
39554.A3
Zinc naphthenate 67% (10% Zn) in Leichtbenzin
Zinc naphthenate
CAS No.: 12001-85-3
Flash Point: 37.8° (100°F)
Physical Form: Liquid
Density: 1.01
Appearance: Colourless
UN No.: UN1993
MDL No.: MFCD00147699