39559.03
39559.06
39559.03EA
79.8
EUR
InStock
39559.03
39559.06
Zinc hexafluoro-2,4-pentanedionate dihydrate
Zinc hexafluoro-2,4-pentanedionate dihydrate
CAS No.: 16743-33-2
Molecular Formula: C10H2F12O4Zn.2H2O
Formula Weight: 515.49 (479.47anhy)
Physical Form: Powder
Melting Point: 157-158°
MDL No.: MFCD00150577