39603.03
39603.06
39603.03EA
49.7
EUR
InStock
39603.03
39603.06
Zinn(II)trifluormethansulfonat ≥97%
Zinn(II)trifluormethansulfonat
Purity: 97%
CAS No.: 62086-04-8
Molecular Formula: Sn(CF3SO3)2
Formula Weight: 416.82
Physical Form: Powder
Sensitivity: Moisture Sensitive
UN No.: 3288
MDL No.: MFCD00191251