39664.30
39664.A1
39664.A7
39664.30EA
72.6
EUR
InStock
39664.30
39664.A1
39664.A7
Titanium(III) sulphate 20% in Schwefelsäure (1 - 4%)
Titanium(III) sulphate
CAS No.: 10343-61-0
Molecular Formula: Ti2(SO4)3
Formula Weight: 383.38
Physical Form: Liquid
Density: 1.15
UN No.: UN3264
MDL No.: MFCD00036317