40030.22
40030.30
40030.36
40030.22EA
69.8
EUR
InStock
40030.22
40030.30
40030.36
Kaliumhexafluorophosphat ≥99%
Potassium hexafluorophosphate
Purity: 99% min
CAS No.: 17084-13-8
Molecular Formula: KPF6
Formula Weight: 184.07
Physical Form: Crystalline
Density: 2.55
Appearance: White
Odour: Odourless
pH: 3.0-7.0
Melting Point: 575°
UN No.: 3260
MDL No.: MFCD00011412