Biotin-11-uridine-5'-triphosphate, tetralithium salt (Biotin-11-UTP) 10 mM in Puffer (Tris-HCl, pH 7.5)
Lieferant: BIOTIUM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
40033
40033.EA 413 EUR InStock
40033.
Biotin-11-uridine-5'-triphosphate, tetralithium salt (Biotin-11-UTP) 10 mM in Puffer (Tris-HCl, pH 7.5)
Biotin-11-uridine-5'-triphosphate, tetralithium salt (Biotin-11-UTP)
Biotin-11-UTP (biotin-11-uridine-5'-triphosphate, tetralithium salt) can be enzymatically incorporated into RNA. The number 11 is the number of atoms in the linker between biotin and UTP.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge