Biotin-20-UTP 10 mM buffer (Tris-HCl, pH 7.5)
Lieferant: BIOTIUM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
40034
40034.EA 498 EUR InStock
40034.
Biotin-20-UTP 10 mM buffer (Tris-HCl, pH 7.5)
Biotin-20-UTP
Biotin-20-UTP can be enzymatically incorporated into RNA. The number 20 is the number of atoms in the linker between biotin and UTP.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge