Fluorescein-12-dUTP, lyophilisiertes Pulver
Lieferant: BIOTIUM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
40063
40063.EA 535 EUR InStock
40063.
Fluorescein-12-dUTP, lyophilisiertes Pulver
Fluorescein-12-dUTP
Fluorescein-12-dUTP can be used to synthesize fluorecently labeled DNA probes for in-situ hybridization, microarray or blotting techniques.
Fluorescein has absorption/emission at 494/521 nm.
Achtung! Bitte beachten: Store Fluorescein-12-dUTP at –20 °C or –70 °C, protected from light.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge