Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Succinimidyl ester methylene blue DNA/RNA Vergleichsfarbstoff

Lieferant: BIOTIUM

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


40075
40075.EA 347 EUR InStock
40075.
Succinimidyl ester methylene blue DNA/RNA Vergleichsfarbstoff
Succinimidyl ester methylene blue
The amine reactive succinimidyl ester of MB (methylene blue) can be conjugated to peptides, proteins, drugs, polymeric materials and biomolecules with primary amine groups. The conjugate will have a blue color and be able to complex with nucleic acids. Methylene blue is a well known redox dye which can be reversibly reduced to its colorless leuco form. Upon oxidation, the blue color of the dye is recovered.

  • Blue solid soluble in DMSO or DMF
  • Absorption 668nm
  • Extinction Coefficient 70,000
  • Stored desiccated at -20°C and protect from light, especially when in solution.


https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd565490b1864b74e8fefff49127c6951ffed5ae&vid=bd565490b1864b74e8fefff49127c6951ffed5ae&prid=web&clid=SAPDAM
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge