unter Argon abgefüllt
40386.04
40386.09
40386.18
40386.04EA
27.8
EUR
InStock
40386.04
40386.09
40386.18
Bis(cyclopentadienyl)titandichlorid ≥99%
Bis(cyclopentadienyl)titandichlorid
Purity: 99+%
CAS No.: 1271-19-8
Molecular Formula: C10H10Cl2Ti
Formula Weight: 249.00
Physical Form: Crystalline, Packaged under argon
Density: 1.60
Appearance: Red
Odour: Odourless
Melting Point: ca 285° dec.
Merck Reference: 14,9482
MDL No.: MFCD00003723