40478.09
40478.22
40478.30
40478.09EA
51.8
EUR
InStock
40478.09
40478.22
40478.30
Niob ≥99,8% (Metall-Basis), Pulver APS 1-5 µm
Niobium
Purity: 99.8%
CAS No.: 7440-03-1
Odour: Odourless
UN No.: 3089
MDL No.: MFCD00011126
Used in high temperature and super alloys of Zn and Ti