40528.14
40528.22
40528.36
40528.14EA
72.7
EUR
InStock
40528.14
40528.22
40528.36
Polyethylenimin, M.W. 1,800 ≥99%, verzweigt
Polyethylenimin
Purity: 99%
CAS No.: 9002-98-6
Molecular Formula: (-NHCH2CH2-)x(-N(CH2CH2NH2)CH2CH2-)y
Physical Form: Liquid
Appearance: Colourless to pale yellow
Odour: Amine-like
MDL No.: MFCD00803910