Mounting media (water-based)
Lieferant: IMMUNOLOGIC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
4058 4059
4058.EA 65.3 EUR InStock
4058. 4059.
Mounting media (water-based)
Färbemittel Mikroskopie- und Färbereagenzien
Imsol Mount is an aqueous mounting medium: Especially for immunohistochemistry slides stained with an alk. phos. method and other non solvent resistant chromogens.
  • Also suitable for immunofluorescence
  • Water based
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge