40980.K2
40980.K2EA
40.4
EUR
InStock
40980.K2
Methanol ≥99,8%, Ökologische Qualität for environmental analysis
Methanol
Purity: 99.8+%
CAS No.: 67-56-1
Molecular Formula: CH3OH
Formula Weight: 32.04
Flash Point: 11°(52°F)
Physical Form: Liquid
Density: 0.791
Refractive Index: 1.3290
Odour: Mild
Melting Point: -98°C
Boiling Point: 64.7
Beilstein Reference: 1098229
Merck Reference: 14,5957
UN No.: 1230
MDL No.: MFCD00004595
Suitable for environmental, pesticide, and GC analysis